Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTGGAAAAAAACTAAGGAAGTTC[C/T]CCACATAACCAGCCGAAGAGGAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603755 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CC2D1B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CC2D1B - coiled-coil and C2 domain containing 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032449.2 | 3924 | UTR 3 | NP_115825.1 | |||
XM_005270590.2 | 3924 | UTR 3 | XP_005270647.1 | |||
XM_005270591.2 | 3924 | Intron | XP_005270648.1 | |||
XM_005270593.2 | 3924 | Intron | XP_005270650.1 | |||
XM_006710423.2 | 3924 | Intron | XP_006710486.1 | |||
XM_017000559.1 | 3924 | Intron | XP_016856048.1 |
ZFYVE9 - zinc finger FYVE-type containing 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |