Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGAGAGACAGGAAATGGGCATCC[A/G]TCAGAAAAACTCGGCAAGAGGAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
18 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf228 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C1orf228 - chromosome 1 open reading frame 228 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145636.1 | Intron | NP_001139108.1 | ||||
XM_006710603.2 | Intron | XP_006710666.1 | ||||
XM_006710604.2 | Intron | XP_006710667.1 | ||||
XM_011541340.1 | Intron | XP_011539642.1 | ||||
XM_011541341.1 | Intron | XP_011539643.1 | ||||
XM_011541343.1 | Intron | XP_011539645.1 | ||||
XM_011541345.1 | Intron | XP_011539647.1 | ||||
XM_011541349.1 | Intron | XP_011539651.1 | ||||
XM_011541354.1 | Intron | XP_011539656.1 | ||||
XM_011541355.1 | Intron | XP_011539657.1 | ||||
XM_011541356.2 | Intron | XP_011539658.1 | ||||
XM_017001140.1 | Intron | XP_016856629.1 | ||||
XM_017001141.1 | Intron | XP_016856630.1 | ||||
XM_017001142.1 | Intron | XP_016856631.1 | ||||
XM_017001143.1 | Intron | XP_016856632.1 | ||||
XM_017001144.1 | Intron | XP_016856633.1 | ||||
XM_017001145.1 | Intron | XP_016856634.1 | ||||
XM_017001146.1 | Intron | XP_016856635.1 | ||||
XM_017001147.1 | Intron | XP_016856636.1 | ||||
XM_017001148.1 | Intron | XP_016856637.1 |