Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGAAAATGAAGTTTCTGTAATGGA[A/G]GCAGGCTTTTTAAGTAGTGACGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614159 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF644 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF644 - zinc finger protein 644 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016620.3 | 572 | Silent Mutation | GCC,GCT | A,A 89 | NP_057704.2 | |
NM_032186.4 | 572 | Silent Mutation | GCC,GCT | A,A 89 | NP_115562.3 | |
NM_201269.2 | 572 | Silent Mutation | GCC,GCT | A,A 1311 | NP_958357.1 | |
XM_005271257.4 | 572 | Silent Mutation | GCC,GCT | A,A 1352 | XP_005271314.1 | |
XM_005271260.4 | 572 | Silent Mutation | GCC,GCT | A,A 1352 | XP_005271317.1 | |
XM_011542258.2 | 572 | Silent Mutation | GCC,GCT | A,A 1352 | XP_011540560.1 | |
XM_011542259.2 | 572 | Silent Mutation | GCC,GCT | A,A 1352 | XP_011540561.1 | |
XM_011542260.2 | 572 | Silent Mutation | GCC,GCT | A,A 1352 | XP_011540562.1 | |
XM_011542261.2 | 572 | Silent Mutation | GCC,GCT | A,A 1352 | XP_011540563.1 | |
XM_017002487.1 | 572 | Silent Mutation | GCC,GCT | A,A 1352 | XP_016857976.1 | |
XM_017002488.1 | 572 | Silent Mutation | GCC,GCT | A,A 1311 | XP_016857977.1 | |
XM_017002489.1 | 572 | Silent Mutation | GCC,GCT | A,A 1311 | XP_016857978.1 | |
XM_017002490.1 | 572 | Silent Mutation | GCC,GCT | A,A 1311 | XP_016857979.1 | |
XM_017002491.1 | 572 | Silent Mutation | GCC,GCT | A,A 1311 | XP_016857980.1 | |
XM_017002492.1 | 572 | Silent Mutation | GCC,GCT | A,A 89 | XP_016857981.1 | |
XM_017002493.1 | 572 | Silent Mutation | GCC,GCT | A,A 89 | XP_016857982.1 | |
XM_017002494.1 | 572 | Silent Mutation | GCC,GCT | A,A 89 | XP_016857983.1 |