Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATAAACCAAAACTGAGGAACTTT[C/G]TACAAACTGGCCAATCCTCCTCAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610496 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP29 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARHGAP29 - Rho GTPase activating protein 29 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004815.3 | 7654 | UTR 3 | NP_004806.3 | |||
XM_006711048.3 | 7654 | Intron | XP_006711111.1 | |||
XM_011542438.2 | 7654 | Intron | XP_011540740.1 | |||
XM_011542439.2 | 7654 | Intron | XP_011540741.1 | |||
XM_011542440.2 | 7654 | Intron | XP_011540742.1 |