Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGATTTCAGACTAAGAAGAGTTT[C/G]GGTACTAAATCTAGGTATTCTGGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614264 MIM: 609607 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP30 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARHGAP30 - Rho GTPase activating protein 30 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NECTIN4 - nectin cell adhesion molecule 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030916.2 | 3302 | UTR 3 | NP_112178.2 | |||
XM_005245508.3 | 3302 | Intron | XP_005245565.1 | |||
XM_011510021.2 | 3302 | Intron | XP_011508323.1 | |||
XM_011510022.2 | 3302 | Intron | XP_011508324.1 | |||
XM_011510023.1 | 3302 | Intron | XP_011508325.1 |