Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGTCATTGTTATTGGGTTCAGTC[A/G]AAATGTATTTGATTACAGGGGATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611471 MIM: 602597 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACP6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACP6 - acid phosphatase 6, lysophosphatidic | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323625.1 | 1888 | Intron | NP_001310554.1 | |||
NM_016361.4 | 1888 | Intron | NP_057445.4 | |||
XM_011509601.2 | 1888 | UTR 3 | XP_011507903.1 | |||
XM_017001422.1 | 1888 | Intron | XP_016856911.1 |
BCL9 - B-cell CLL/lymphoma 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |