Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTCGTCTTCTCTCTCCTCGATTG[C/T]TGCGCGCTCATCTTCCTCTCGGTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
14 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNIH4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CNIH4 - cornichon family AMPA receptor auxiliary protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277197.1 | 153 | Silent Mutation | TGC,TGT | C,C 13 | NP_001264126.1 | |
NM_001277198.1 | 153 | Silent Mutation | TGC,TGT | C,C 13 | NP_001264127.1 | |
NM_001277199.1 | 153 | UTR 5 | NP_001264128.1 | |||
NM_001277200.1 | 153 | Silent Mutation | TGC,TGT | C,C 13 | NP_001264129.1 | |
NM_014184.3 | 153 | Silent Mutation | TGC,TGT | C,C 13 | NP_054903.1 |