Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGACGCGGGTTTCTCAGACATCG[C/T]GTCCTCAGTGCCCTTGCCAGTCTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
8 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612532 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PHF13 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PHF13 - PHD finger protein 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153812.2 | 604 | Missense Mutation | GCG,GTG | A,V 78 | NP_722519.2 | |
XM_011540762.1 | 604 | Missense Mutation | GCG,GTG | A,V 87 | XP_011539064.1 |
THAP3 - THAP domain containing 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |