Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTTGTTGCTGTCAGTGGCTGGCA[A/G]CCGCTTGTAGTTGAACAGGCCTTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
8 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608867 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DUSP10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DUSP10 - dual specificity phosphatase 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007207.5 | 1356 | Silent Mutation | CTG,TTG | L,L 373 | NP_009138.1 | |
XM_017000147.1 | 1356 | Intron | XP_016855636.1 |