Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGAAATGCTGAGAATGCCAGGCA[C/T]CCCAGTTCTGAGAGCCACCCTAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603155 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTPN14 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PTPN14 - protein tyrosine phosphatase, non-receptor type 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005401.4 | 12202 | UTR 3 | NP_005392.2 | |||
XM_017001941.1 | 12202 | Intron | XP_016857430.1 |