Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGACATATTAAGGCAACCAGCCA[C/T]GCCATCTGGCCAGCACAAGGGACTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616262 MIM: 165270 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLHL21 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KLHL21 - kelch like family member 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324309.1 | 3121 | UTR 3 | NP_001311238.1 | |||
NM_014851.3 | 3121 | UTR 3 | NP_055666.2 |
ZBTB48 - zinc finger and BTB domain containing 48 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |