Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTGTGTATAGTTGCTGCTTCAGA[A/G]TCTGGTATTAAGAGGCTGAAATCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606511 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MARK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MARK1 - microtubule affinity regulating kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286124.1 | 404 | Intron | NP_001273053.1 | |||
NM_001286126.1 | 404 | Intron | NP_001273055.1 | |||
NM_001286128.1 | 404 | Intron | NP_001273057.1 | |||
NM_001286129.1 | 404 | Intron | NP_001273058.1 | |||
NM_018650.4 | 404 | Intron | NP_061120.3 | |||
XM_005273134.4 | 404 | Intron | XP_005273191.1 | |||
XM_006711326.3 | 404 | Intron | XP_006711389.1 | |||
XM_011509561.2 | 404 | UTR 5 | XP_011507863.1 | |||
XM_011509562.2 | 404 | Intron | XP_011507864.1 | |||
XM_017001305.1 | 404 | Intron | XP_016856794.1 | |||
XM_017001306.1 | 404 | Intron | XP_016856795.1 | |||
XM_017001307.1 | 404 | Intron | XP_016856796.1 |