Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATAAAAACAAATGAAGTACAAAAT[A/G]TTTCAGATTTACATAGTGATAAACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608690 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SSX2IP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SSX2IP - SSX family member 2 interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166293.1 | 4490 | UTR 3 | NP_001159765.1 | |||
NM_001166294.1 | 4490 | UTR 3 | NP_001159766.1 | |||
NM_001166295.1 | 4490 | UTR 3 | NP_001159767.1 | |||
NM_001166417.1 | 4490 | UTR 3 | NP_001159889.1 | |||
NM_014021.3 | 4490 | UTR 3 | NP_054740.3 | |||
XM_005270427.1 | 4490 | UTR 3 | XP_005270484.1 | |||
XM_005270428.1 | 4490 | UTR 3 | XP_005270485.1 | |||
XM_005270429.1 | 4490 | UTR 3 | XP_005270486.1 | |||
XM_006710331.1 | 4490 | UTR 3 | XP_006710394.1 | |||
XM_011540610.1 | 4490 | UTR 3 | XP_011538912.1 | |||
XM_011540612.2 | 4490 | Intron | XP_011538914.1 | |||
XM_017000234.1 | 4490 | UTR 3 | XP_016855723.1 | |||
XM_017000235.1 | 4490 | UTR 3 | XP_016855724.1 | |||
XM_017000236.1 | 4490 | UTR 3 | XP_016855725.1 | |||
XM_017000237.1 | 4490 | UTR 3 | XP_016855726.1 |