Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGAGGGGGCACTGCTCCCTAGTCT[A/C]TGTCACCCAGGGTTTACCTCCCCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
17 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610278 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PEAR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PEAR1 - platelet endothelial aggregation receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080471.1 | Intron | NP_001073940.1 | ||||
XM_005245141.3 | Intron | XP_005245198.1 | ||||
XM_011509510.2 | Intron | XP_011507812.1 | ||||
XM_011509511.2 | Intron | XP_011507813.1 | ||||
XM_011509512.2 | Intron | XP_011507814.1 | ||||
XM_017001234.1 | Intron | XP_016856723.1 | ||||
XM_017001235.1 | Intron | XP_016856724.1 | ||||
XM_017001236.1 | Intron | XP_016856725.1 | ||||
XM_017001237.1 | Intron | XP_016856726.1 | ||||
XM_017001238.1 | Intron | XP_016856727.1 | ||||
XM_017001239.1 | Intron | XP_016856728.1 | ||||
XM_017001240.1 | Intron | XP_016856729.1 | ||||
XM_017001241.1 | Intron | XP_016856730.1 | ||||
XM_017001242.1 | Intron | XP_016856731.1 |