Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGGACTGGTTCTTTGAGGTTCTG[C/T]CCTTTTTCCATGATTCAGACCAACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
14 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616119 MIM: 602413 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CFAP126 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CFAP126 - cilia and flagella associated protein 126 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013625.3 | 2492 | Intron | NP_001013647.2 |
SDHC - succinate dehydrogenase complex subunit C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001035511.1 | 2492 | UTR 3 | NP_001030588.1 | |||
NM_001035512.1 | 2492 | UTR 3 | NP_001030589.1 | |||
NM_001035513.1 | 2492 | UTR 3 | NP_001030590.1 | |||
NM_001278172.1 | 2492 | UTR 3 | NP_001265101.1 | |||
NM_003001.3 | 2492 | UTR 3 | NP_002992.1 |