Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCCCAGCCCTAAAGGTTGTGGCT[A/G]CATCCTGCAGTTATATTAATACCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600904 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASTN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ASTN1 - astrotactin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286164.1 | 4982 | Intron | NP_001273093.1 | |||
NM_004319.2 | 4982 | Intron | NP_004310.1 | |||
NM_207108.2 | 4982 | Intron | NP_996991.1 | |||
XM_017001340.1 | 4982 | Intron | XP_016856829.1 | |||
XM_017001341.1 | 4982 | UTR 3 | XP_016856830.1 |