Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAGCTGGTCACTAACTTTGCAGA[C/T]GGATGAGCCTTGAGCACCCAGAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605254 MIM: 162360 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NCSTN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NCSTN - nicastrin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NHLH1 - nescient helix-loop-helix 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005598.3 | 178 | UTR 5 | NP_005589.1 |