Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGGGCTTCTGGGTCAGAGGATCG[C/T]CCGCCTGTTGGTGGAAGAGAAGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613890 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSD3B2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HSD3B2 - hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000198.3 | 305 | Missense Mutation | GCC,GTC | A,V 19 | NP_000189.1 | |
NM_001166120.1 | 305 | Missense Mutation | GCC,GTC | A,V 19 | NP_001159592.1 |