Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGCGTCAAGGTGAACGAGGCCTAC[C/G]GGTTCCGCGTGGCACTGCCTGCGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
26 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600347 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCAN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BCAN - brevican | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021948.4 | 649 | Missense Mutation | CGG,GGG | R,G 105 | NP_068767.3 | |
NM_198427.1 | 649 | Missense Mutation | CGG,GGG | R,G 105 | NP_940819.1 | |
XM_011509866.1 | 649 | Missense Mutation | CGG,GGG | R,G 105 | XP_011508168.1 | |
XM_017002047.1 | 649 | Missense Mutation | CGG,GGG | R,G 150 | XP_016857536.1 |