Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAAAGGTCTACCCCTTACCCATGA[C/T]ATTGGTGTTACTATTTCTTTGGGTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
SLC9C2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC9C2 - solute carrier family 9 member C2 (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178527.3 | 3607 | Missense Mutation | ATC,GTC | I,V 1102 | NP_848622.2 | |
XM_011509426.1 | 3607 | Missense Mutation | ATC,GTC | I,V 1102 | XP_011507728.1 | |
XM_017001065.1 | 3607 | Missense Mutation | ATC,GTC | I,V 1102 | XP_016856554.1 | |
XM_017001066.1 | 3607 | Missense Mutation | ATC,GTC | I,V 1102 | XP_016856555.1 | |
XM_017001067.1 | 3607 | Missense Mutation | ATC,GTC | I,V 1062 | XP_016856556.1 | |
XM_017001068.1 | 3607 | Missense Mutation | ATC,GTC | I,V 1060 | XP_016856557.1 | |
XM_017001069.1 | 3607 | Missense Mutation | ATC,GTC | I,V 1060 | XP_016856558.1 | |
XM_017001070.1 | 3607 | Missense Mutation | ATC,GTC | I,V 1060 | XP_016856559.1 | |
XM_017001071.1 | 3607 | Missense Mutation | ATC,GTC | I,V 1056 | XP_016856560.1 | |
XM_017001072.1 | 3607 | Missense Mutation | ATC,GTC | I,V 1013 | XP_016856561.1 | |
XM_017001073.1 | 3607 | Missense Mutation | ATC,GTC | I,V 1000 | XP_016856562.1 | |
XM_017001074.1 | 3607 | Missense Mutation | ATC,GTC | I,V 1000 | XP_016856563.1 | |
XM_017001075.1 | 3607 | Missense Mutation | ATC,GTC | I,V 742 | XP_016856564.1 | |
XM_017001076.1 | 3607 | Missense Mutation | ATC,GTC | I,V 660 | XP_016856565.1 |