Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCTAACAGGTGGAGTTAGAAAAG[C/T]GGGTGCCACAAACTATGGTTTAATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608917 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATPAF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATPAF1 - ATP synthase mitochondrial F1 complex assembly factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042546.2 | 1692 | UTR 3 | NP_001036011.2 | |||
NM_001243728.1 | 1692 | UTR 3 | NP_001230657.1 | |||
NM_001256418.1 | 1692 | UTR 3 | NP_001243347.1 | |||
NM_022745.4 | 1692 | UTR 3 | NP_073582.3 |