Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATGTAATGGTGTTATCATTGGAT[A/G]GGAGGAATTATACCAAGTTTCTCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602162 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SYCP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SYCP1 - synaptonemal complex protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282541.1 | Intron | NP_001269470.1 | ||||
NM_001282542.1 | Intron | NP_001269471.1 | ||||
NM_003176.3 | Intron | NP_003167.2 | ||||
XM_005271155.3 | Intron | XP_005271212.1 | ||||
XM_005271156.1 | Intron | XP_005271213.1 | ||||
XM_006710859.1 | Intron | XP_006710922.1 | ||||
XM_011542037.2 | Intron | XP_011540339.1 | ||||
XM_011542038.2 | Intron | XP_011540340.1 | ||||
XM_011542039.1 | Intron | XP_011540341.1 | ||||
XM_017002184.1 | Intron | XP_016857673.1 | ||||
XM_017002185.1 | Intron | XP_016857674.1 |