Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATCCTGTGGTGACTGCCATCTTG[A/C]ATCACTGTTGCCCCATGTTTTGTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
14 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600236 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CENPF PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CENPF - centromere protein F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016343.3 | Intron | NP_057427.3 | ||||
XM_011509082.2 | Intron | XP_011507384.1 | ||||
XM_017000086.1 | Intron | XP_016855575.1 |