Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTAAAGACATAACTCATATCAGTC[A/G]CATCATTGGACCCATCCACACCTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602715 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IPO9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IPO9 - importin 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LMOD1 - leiomodin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SHISA4 - shisa family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198149.2 | 1120 | UTR 3 | NP_937792.2 | |||
XM_017000419.1 | 1120 | UTR 3 | XP_016855908.1 |