Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAATGGAGTTGAGTGTATTTTTA[C/T]TTAAATGACTTATAGTATTTTACTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR4742 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR4742 - microRNA 4742 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WDR26 - WD repeat domain 26 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001115113.2 | Intron | NP_001108585.2 | ||||
NM_025160.6 | Intron | NP_079436.4 | ||||
XM_005273273.4 | Intron | XP_005273330.1 | ||||
XM_006711817.3 | Intron | XP_006711880.1 | ||||
XM_011544281.2 | Intron | XP_011542583.1 | ||||
XM_011544282.2 | Intron | XP_011542584.1 | ||||
XM_017002396.1 | Intron | XP_016857885.1 |