Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTCGGCTTCTGCACGGATACACT[A/G]TGTTGTGCCATCCAACTGGAGTGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
19 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TTC39A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TTC39A - tetratricopeptide repeat domain 39A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080494.3 | 2074 | UTR 3 | NP_001073963.1 | |||
NM_001144832.2 | 2074 | UTR 3 | NP_001138304.1 | |||
NM_001297662.1 | 2074 | Intron | NP_001284591.1 | |||
NM_001297663.1 | 2074 | UTR 3 | NP_001284592.1 | |||
NM_001297664.1 | 2074 | UTR 3 | NP_001284593.1 | |||
NM_001297665.1 | 2074 | UTR 3 | NP_001284594.1 | |||
NM_001297666.1 | 2074 | UTR 3 | NP_001284595.1 | |||
NM_001297667.1 | 2074 | UTR 3 | NP_001284596.1 | |||
XM_005270644.2 | 2074 | UTR 3 | XP_005270701.1 | |||
XM_006710471.2 | 2074 | UTR 3 | XP_006710534.1 | |||
XM_011541048.2 | 2074 | UTR 3 | XP_011539350.1 | |||
XM_011541049.2 | 2074 | UTR 3 | XP_011539351.1 | |||
XM_011541050.2 | 2074 | UTR 3 | XP_011539352.1 | |||
XM_011541051.2 | 2074 | UTR 3 | XP_011539353.1 | |||
XM_011541052.2 | 2074 | UTR 3 | XP_011539354.1 | |||
XM_011541053.2 | 2074 | UTR 3 | XP_011539355.1 | |||
XM_011541054.2 | 2074 | UTR 3 | XP_011539356.1 | |||
XM_017000715.1 | 2074 | UTR 3 | XP_016856204.1 |