Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATGTCTTCTGGGGGGAAAGCCCC[A/C]ACCTCCACGATCTCTTCCACCTCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
8 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602284 MIM: 610289 MIM: 602435 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BMP8B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BMP8B - bone morphogenetic protein 8b | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005271149.4 | Intron | XP_005271206.1 | ||||
XM_011542022.2 | Intron | XP_011540324.1 | ||||
XM_011542024.2 | Intron | XP_011540326.2 | ||||
XM_011542025.2 | Intron | XP_011540327.1 | ||||
XM_017002155.1 | Intron | XP_016857644.1 | ||||
XM_017002156.1 | Intron | XP_016857645.1 |
OXCT2 - 3-oxoacid CoA-transferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022120.1 | Intron | NP_071403.1 |
PPIE - peptidylprolyl isomerase E | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |