Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGAGGCCCTTGAACTTTTAGGTT[A/T]TGCTCTGAGGCTGTGGCAACCAACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
18 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605995 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIF1B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIF1B - kinesin family member 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015074.3 | Intron | NP_055889.2 | ||||
NM_183416.3 | Intron | NP_904325.2 |
MIR1273D - microRNA 1273d | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |