Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAACCCAACAATATTAGATCAATA[C/T]GTAGTATTTAAGTGTCCATAATGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615014 MIM: 602797 MIM: 601831 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HIST2H2AB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HIST2H2AB - histone cluster 2, H2ab | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HIST2H2AC - histone cluster 2, H2ac | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HIST2H2BE - histone cluster 2, H2be | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003528.2 | 1420 | UTR 3 | NP_003519.1 |