Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAAACTTATTAAATTGACTCTTAA[A/G]CTAAGTTTTTAGTCTTTAATTTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 111700 MIM: 605348 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RHCE PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RHCE - Rh blood group CcEe antigens | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020485.4 | 1376 | UTR 3 | NP_065231.3 | |||
NM_138616.3 | 1376 | UTR 3 | NP_619522.3 | |||
NM_138617.3 | 1376 | UTR 3 | NP_619523.3 | |||
NM_138618.3 | 1376 | UTR 3 | NP_619524.3 | |||
XM_005245957.3 | 1376 | Intron | XP_005246014.1 | |||
XM_006710810.3 | 1376 | Intron | XP_006710873.1 | |||
XM_011541888.2 | 1376 | Intron | XP_011540190.1 | |||
XM_011541889.2 | 1376 | Intron | XP_011540191.1 | |||
XM_011541890.2 | 1376 | Intron | XP_011540192.1 | |||
XM_011541891.2 | 1376 | Intron | XP_011540193.1 | |||
XM_017002014.1 | 1376 | Intron | XP_016857503.1 |
TMEM50A - transmembrane protein 50A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014313.3 | 1376 | UTR 3 | NP_055128.1 | |||
XM_005245817.1 | 1376 | UTR 3 | XP_005245874.1 | |||
XM_011541159.1 | 1376 | UTR 3 | XP_011539461.1 |