Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGGGTGGGCGGGGCCCGGTGGGC[A/G]GGGCCCTATCAGTGCTGGGCTCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
14 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142461 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSPG2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HSPG2 - heparan sulfate proteoglycan 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291860.1 | 13992 | UTR 3 | NP_001278789.1 | |||
NM_005529.6 | 13992 | UTR 3 | NP_005520.4 | |||
XM_011541318.2 | 13992 | UTR 3 | XP_011539620.1 | |||
XM_017001120.1 | 13992 | UTR 3 | XP_016856609.1 | |||
XM_017001121.1 | 13992 | UTR 3 | XP_016856610.1 | |||
XM_017001122.1 | 13992 | UTR 3 | XP_016856611.1 |
LDLRAD2 - low density lipoprotein receptor class A domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013693.2 | 13992 | UTR 3 | NP_001013715.2 | |||
XM_005245873.4 | 13992 | Intron | XP_005245930.1 |