Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTTGTAGATGTTTTCTTTTCTGG[C/T]GGGCTTTCCGGTTCCTCATCTTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611912 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUCKS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NUCKS1 - nuclear casein kinase and cyclin dependent kinase substrate 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022731.4 | 889 | Silent Mutation | NP_073568.2 | |||
XM_005245453.1 | 889 | Silent Mutation | XP_005245510.1 |