Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACTAATGTGAGCCAGCAACATGT[C/G]AAAGGCTAATCCAGGAGATGCAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
8 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603949 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAB29 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RAB29 - RAB29, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135662.1 | 750 | UTR 3 | NP_001129134.1 | |||
NM_001135663.1 | 750 | UTR 3 | NP_001129135.1 | |||
NM_001135664.1 | 750 | UTR 3 | NP_001129136.1 | |||
NM_003929.2 | 750 | UTR 3 | NP_003920.1 | |||
XM_005245569.1 | 750 | UTR 3 | XP_005245626.1 | |||
XM_005245570.1 | 750 | UTR 3 | XP_005245627.1 | |||
XM_005245571.1 | 750 | UTR 3 | XP_005245628.1 | |||
XM_006711605.3 | 750 | UTR 3 | XP_006711668.1 | |||
XM_006711606.2 | 750 | UTR 3 | XP_006711669.1 | |||
XM_017002748.1 | 750 | UTR 3 | XP_016858237.1 | |||
XM_017002749.1 | 750 | UTR 3 | XP_016858238.1 | |||
XM_017002750.1 | 750 | UTR 3 | XP_016858239.1 |