Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAGAGGAGGAAGGCATGGGAGGG[A/G]CATCAGCTGTGTCTGGAGGATGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
18 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610816 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC25A33 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC25A33 - solute carrier family 25 member 33 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM201 - transmembrane protein 201 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010866.3 | Intron | NP_001010866.1 | ||||
NM_001130924.2 | Intron | NP_001124396.2 | ||||
XM_006710417.3 | Intron | XP_006710480.1 | ||||
XM_011540910.2 | Intron | XP_011539212.1 | ||||
XM_017000549.1 | Intron | XP_016856038.1 | ||||
XM_017000550.1 | Intron | XP_016856039.1 | ||||
XM_017000551.1 | Intron | XP_016856040.1 | ||||
XM_017000552.1 | Intron | XP_016856041.1 |