Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCTGCAGTGAGCCATATCCTGTT[A/C]ATCTCTGTATCCTTGGATCCTAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606095 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPIH PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPIH - peptidylprolyl isomerase H | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006347.3 | Intron | NP_006338.1 | ||||
XM_005270362.1 | Intron | XP_005270419.1 | ||||
XM_005270366.3 | Intron | XP_005270423.1 | ||||
XM_011540508.1 | Intron | XP_011538810.1 | ||||
XM_017000057.1 | Intron | XP_016855546.1 | ||||
XM_017000058.1 | Intron | XP_016855547.1 | ||||
XM_017000059.1 | Intron | XP_016855548.1 |