Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTTTCGAGGCTGCAGAAGCACCC[C/T]CAGGGCCACAAAGCCCCATCTGTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605492 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105371692 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105371692 - uncharacterized LOC105371692 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRRN2 - leucine rich repeat neuronal 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006338.2 | 2756 | UTR 3 | NP_006329.2 | |||
NM_201630.1 | 2756 | UTR 3 | NP_963924.1 | |||
XM_005244827.3 | 2756 | UTR 3 | XP_005244884.1 | |||
XM_011509066.2 | 2756 | UTR 3 | XP_011507368.1 | |||
XM_017000050.1 | 2756 | UTR 3 | XP_016855539.1 |