Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACAATGTTATAAAACAAAAGCAA[A/C]GTATACACACTCTATACACACGTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612666 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DSTYK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DSTYK - dual serine/threonine and tyrosine protein kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015375.2 | 5769 | UTR 3 | NP_056190.1 | |||
NM_199462.2 | 5769 | UTR 3 | NP_955749.1 | |||
XM_011509392.2 | 5769 | UTR 3 | XP_011507694.1 | |||
XM_011509393.2 | 5769 | UTR 3 | XP_011507695.1 | |||
XM_011509394.2 | 5769 | UTR 3 | XP_011507696.1 |