Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTATTCTTCCCTTGCATTCTTCAG[C/T]GGTCTCAGCTCCTTTCTCTGTGTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606524 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SRGAP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SRGAP2 - SLIT-ROBO Rho GTPase activating protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170637.3 | Intron | NP_001164108.1 | ||||
NM_001300952.1 | Intron | NP_001287881.1 | ||||
NM_015326.4 | Intron | NP_056141.2 | ||||
XM_005277510.2 | Intron | XP_005277567.1 | ||||
XM_005277511.2 | Intron | XP_005277568.1 | ||||
XM_005277514.1 | Intron | XP_005277571.1 | ||||
XM_005277515.1 | Intron | XP_005277572.1 | ||||
XM_011509354.1 | Intron | XP_011507656.1 | ||||
XM_011509355.1 | Intron | XP_011507657.1 | ||||
XM_011509356.2 | Intron | XP_011507658.1 | ||||
XM_011509359.1 | Intron | XP_011507661.1 | ||||
XM_017000838.1 | Intron | XP_016856327.1 | ||||
XM_017000839.1 | Intron | XP_016856328.1 | ||||
XM_017000840.1 | Intron | XP_016856329.1 | ||||
XM_017000841.1 | Intron | XP_016856330.1 |