Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCTGCTGACCTCCGAGCGCACAG[A/G]CGCACGCATGCTGGCCAGACCCTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF672 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF672 - zinc finger protein 672 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024836.2 | 1031 | Silent Mutation | AGA,AGG | R,R 61 | NP_079112.1 | |
XM_005270336.1 | 1031 | Silent Mutation | AGA,AGG | R,R 61 | XP_005270393.1 |
ZNF692 - zinc finger protein 692 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |