Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAACAAAATTATGGGAGAGTTCTGA[A/C]ATTTCCGTAGATTCTGCAAATTTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 609448 | ||||||||||||||||||||
Literature Links: |
KTI12 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KTI12 - KTI12 chromatin associated homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TXNDC12 - thioredoxin domain containing 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015913.3 | 1321 | Missense Mutation | ATG,ATT | M,I 82 | NP_056997.1 |