Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTGGTTCATGCCATCTGCCTGTT[A/C]GGGGAATACGCAGACTCACACTATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138380 MIM: 138333 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GSTM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GSTM2 - glutathione S-transferase mu 2 (muscle) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GSTM4 - glutathione S-transferase mu 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000850.4 | Intron | NP_000841.1 | ||||
NM_147148.2 | Intron | NP_671489.1 | ||||
XM_017001085.1 | Intron | XP_016856574.1 | ||||
XM_017001086.1 | Intron | XP_016856575.1 | ||||
XM_017001087.1 | Intron | XP_016856576.1 | ||||
XM_017001088.1 | Intron | XP_016856577.1 |