Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACCCCCTCTACGCTCTCCTTCCT[C/T]GCCAGCACGCCTTAGCTTTGCAAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614981 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATPIF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATPIF1 - ATPase inhibitory factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016311.4 | 700 | Intron | NP_057395.1 | |||
NM_178190.2 | 700 | Intron | NP_835497.1 | |||
NM_178191.2 | 700 | UTR 3 | NP_835498.1 |
DNAJC8 - DnaJ heat shock protein family (Hsp40) member C8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105376896 - uncharacterized LOC105376896 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |