Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCTGGGGTAGATGGAAGGGCCAC[A/G]TCTGTCCAGCCATCGCCCTGGCAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600997 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EPHB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EPHB2 - EPH receptor B2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001309192.1 | Intron | NP_001296121.1 | ||||
NM_001309193.1 | Intron | NP_001296122.1 | ||||
NM_004442.7 | Intron | NP_004433.2 | ||||
NM_017449.4 | Intron | NP_059145.2 | ||||
XM_006710441.3 | Intron | XP_006710504.1 | ||||
XM_006710442.3 | Intron | XP_006710505.1 |
MIR4684 - microRNA 4684 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |