Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTACAAAATATTAAAAAGTCAGT[C/T]AATCATCTACACAGTACCCCCCATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611321 MIM: 602839 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLSTN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLSTN1 - calsyntenin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009566.2 | 4220 | UTR 3 | NP_001009566.1 | |||
NM_001302883.1 | 4220 | UTR 3 | NP_001289812.1 | |||
NM_014944.4 | 4220 | UTR 3 | NP_055759.3 |
PIK3CD - phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit delta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005026.3 | 4220 | Intron | NP_005017.3 | |||
XM_006710687.2 | 4220 | Intron | XP_006710750.1 | |||
XM_006710689.2 | 4220 | Intron | XP_006710752.1 | |||
XM_017001476.1 | 4220 | Intron | XP_016856965.1 | |||
XM_017001477.1 | 4220 | Intron | XP_016856966.1 | |||
XM_017001478.1 | 4220 | Intron | XP_016856967.1 | |||
XM_017001479.1 | 4220 | Intron | XP_016856968.1 | |||
XM_017001480.1 | 4220 | Intron | XP_016856969.1 | |||
XM_017001481.1 | 4220 | Intron | XP_016856970.1 | |||
XM_017001482.1 | 4220 | Intron | XP_016856971.1 | |||
XM_017001483.1 | 4220 | Intron | XP_016856972.1 | |||
XM_017001484.1 | 4220 | Intron | XP_016856973.1 | |||
XM_017001485.1 | 4220 | Intron | XP_016856974.1 |