Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATATGTGTTTGCCTGCTTTTACTA[G/T]ACTACATATGTACATATTTACATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
8 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLPPR5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PLPPR5 - phospholipid phosphatase related 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010861.2 | 4102 | UTR 3 | NP_001010861.1 | |||
NM_001037317.1 | 4102 | UTR 3 | NP_001032394.1 | |||
XM_011540836.2 | 4102 | UTR 3 | XP_011539138.1 | |||
XM_011540837.1 | 4102 | Intron | XP_011539139.1 | |||
XM_011540838.2 | 4102 | UTR 3 | XP_011539140.1 |