Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCACGGGGCCGGTAATTTGGTCA[A/G]CACTTACTATGACTTTTCCAAGGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 611632 | ||||||||||||||||||||
Literature Links: |
UBIAD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
UBIAD1 - UbiA prenyltransferase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013319.2 | 639 | Missense Mutation | AAC,AGC | N,S 102 | NP_037451.1 | |
XM_006710590.3 | 639 | Missense Mutation | AAC,AGC | N,S 102 | XP_006710653.1 | |
XM_011541304.2 | 639 | Missense Mutation | AAC,AGC | N,S 102 | XP_011539606.1 |