Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGAACCAGATAGAGCCCGGCGTGC[A/G]GTACAACTACGTGTACGACGAGGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 102573 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACTN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACTN2 - actinin alpha 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001103.3 | 246 | Missense Mutation | CAG,CGG | Q,R 9 | NP_001094.1 | |
NM_001278343.1 | 246 | Missense Mutation | CAG,CGG | Q,R 9 | NP_001265272.1 | |
NM_001278344.1 | 246 | UTR 5 | NP_001265273.1 |