Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTGTAGCACCATCAATAACTCTG[T/C]GATCAGCTGACCAGCTCACATTCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 248610 | ||||||||||||||||||||
Literature Links: |
DBT PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DBT - dihydrolipoamide branched chain transacylase E2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001918.3 | 1615 | Missense Mutation | CAC,CGC | H,R 452 | NP_001909.3 | |
XM_005270545.3 | 1615 | Missense Mutation | CAC,CGC | H,R 271 | XP_005270602.1 | |
XM_017000468.1 | 1615 | Missense Mutation | CAC,CGC | H,R 271 | XP_016855957.1 | |
XM_017000469.1 | 1615 | Missense Mutation | CAC,CGC | H,R 271 | XP_016855958.1 |