Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAACGCTACAACGCGGACCGTCTG[C/T]GTCACGGCCCCATAGGCCTCCCGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607622 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PMVK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PMVK - phosphomevalonate kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323011.1 | 701 | Silent Mutation | ACA,ACG | T,T 112 | NP_001309940.1 | |
NM_001323012.1 | 701 | Silent Mutation | ACA,ACG | T,T 51 | NP_001309941.1 | |
NM_006556.3 | 701 | Silent Mutation | ACA,ACG | T,T 126 | NP_006547.1 |