Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCCAGGAAGTGCAGGGCAGTGGC[A/G]CTTTGATCATACACAGCAAACAGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 106150 MIM: 606974 | ||||||||||||||||||||
Literature Links: |
AGT PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AGT - angiotensinogen | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000029.3 | 1912 | Silent Mutation | AGC,AGT | S,S 468 | NP_000020.1 |
COG2 - component of oligomeric golgi complex 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |